Assay Details
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005601087.3 | XP_005601144.1 | 484 | 75 | ||
XM_023621362.1 | XP_023477130.1 | 847 | 75 | |||
XM_023621363.1 | XP_023477131.1 | 847 | 75 | |||
XM_023621364.1 | XP_023477132.1 | 632 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
