Assay Details
Gene Symbol: LATS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621368.1 | XP_023477136.1 | 2895 | 67 | ||
XM_023621369.1 | XP_023477137.1 | 3370 | 67 | |||
XM_023621370.1 | XP_023477138.1 | 2891 | 67 | |||
XM_023621371.1 | XP_023477139.1 | 5239 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
