Assay Details
Gene Symbol: C1QTNF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490075.4 | XP_001490125.1 | 335 | 79 | ||
XM_023621393.1 | XP_023477161.1 | 335 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
