Assay Details
Gene Symbol: GTF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621335.1 | XP_023477103.1 | 552 | 98 | ||
GenBank mRNA | HQ890201.1 | - | 1 - 2 | 78 | 98 | |
JL623043.1 | - | 2 - 3 | 138 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
