Assay Details
Gene Symbol: MTERF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610763.3 | XP_005610820.1 | 318 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005610763.3 transcripts.
Set Membership: |
![]() ![]() |
