Assay Details
Gene Symbol: MTERF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497592.4 | XP_001497642.1 | 794 | 71 | ||
XM_005610762.3 | XP_005610819.1 | 1187 | 71 | |||
XM_005610763.3 | XP_005610820.1 | 993 | 71 | |||
XM_023644068.1 | XP_023499836.1 | 805 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
