Assay Details
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023642795.1 | XP_023498563.1 | 661 | 64 | ||
XM_023642797.1 | XP_023498565.1 | 404 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
