Assay Details
Gene Symbol: DTYMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497782.4 | XP_001497832.4 | 748 | 65 | ||
XM_014740419.2 | XP_014595905.2 | 748 | 65 | |||
XM_023642804.1 | XP_023498572.1 | 301 | 65 | |||
XM_023642805.1 | XP_023498573.1 | 295 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
