Assay Details
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610818.3 | XP_005610875.1 | 589 | 74 | ||
XM_023642857.1 | XP_023498625.1 | 631 | 74 | |||
XM_023642858.1 | XP_023498626.1 | 567 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
