Assay Details
Gene Symbol: HDHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610823.3 | XP_005610880.1 | 812 | 60 | ||
XM_023642868.1 | XP_023498636.1 | 819 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
