Assay Details
Gene Symbol: IQSEC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023642969.1 | XP_023498737.1 | 2181 | 62 | ||
XM_023642970.1 | XP_023498738.1 | 2181 | 62 | |||
XM_023642971.1 | XP_023498739.1 | 2181 | 62 | |||
XM_023642972.1 | XP_023498740.1 | 2181 | 62 | |||
XM_023642973.1 | XP_023498741.1 | 2181 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
