Assay Details
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610847.3 | XP_005610904.1 | 429 | 72 | ||
XM_023642992.1 | XP_023498760.1 | 1128 | 72 | |||
XM_023642994.1 | XP_023498762.1 | 288 | 72 | |||
XM_023642995.1 | XP_023498763.1 | 364 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
