Assay Details
Gene Symbol: CD27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610858.3 | XP_005610915.1 | 1046 | 66 | ||
XM_005610859.3 | XP_005610916.1 | 920 | 66 | |||
XM_023643026.1 | XP_023498794.1 | 967 | 66 | |||
XM_023643027.1 | XP_023498795.1 | 878 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
