Assay Details
Gene Symbol: SPART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495257.4 | XP_001495307.1 | 2038 | 90 | ||
XM_005601141.3 | XP_005601198.1 | 2597 | 90 | |||
XM_005601143.3 | XP_005601200.1 | 1941 | 90 | |||
XM_005601144.3 | XP_005601201.1 | 2111 | 90 | |||
GenBank mRNA | JL638549.1 | - | 7 - 8 | 1058 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
