Assay Details
Gene Symbol: ALG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496358.6 | XP_001496408.2 | 965 | 69 | ||
XM_005601149.3 | XP_005601206.1 | 872 | 69 | |||
XM_005601150.3 | XP_005601207.1 | 851 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
