Assay Details
Gene Symbol: MRPS31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005601180.3 | XP_005601237.1 | 913 | 62 | ||
XM_014732060.2 | XP_014587546.1 | 926 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
