Assay Details
Gene Symbol: SPRYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014732077.2 | XP_014587563.2 | 838 | 96 | ||
GenBank mRNA | JL625029.1 | - | 1 - 2 | 171 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
