Assay Details
Gene Symbol: ESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490196.5 | XP_001490246.1 | 402 | 65 | ||
XM_005601225.3 | XP_005601282.1 | 560 | 65 | |||
XM_005601226.3 | XP_005601283.1 | 403 | 65 | |||
XM_023621595.1 | XP_023477363.1 | 2166 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
