Assay Details
Gene Symbol: RGCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493104.5 | XP_001493154.1 | 837 | 83 | ||
XM_023621653.1 | XP_023477421.1 | 473 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
