Assay Details
Gene Symbol: MTRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492709.4 | XP_001492759.1 | 1735 | 61 | ||
XM_005601259.3 | XP_005601316.1 | 1856 | 61 | |||
XM_014732118.2 | XP_014587604.1 | 1582 | 61 | |||
XM_014732119.2 | XP_014587605.1 | 1468 | 61 | |||
XM_023621660.1 | XP_023477428.1 | 1853 | 61 | |||
XM_023621661.1 | XP_023477429.1 | 1706 | 61 | |||
GenBank mRNA | JL625825.1 | - | 7 - 8 | 886 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
