Assay Details
Gene Symbol: CNMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493004.4 | XP_001493054.1 | 186 | 92 | ||
XM_005601262.3 | XP_005601319.1 | 188 | 92 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CNMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493004.4 | XP_001493054.1 | 186 | 92 | ||
XM_005601262.3 | XP_005601319.1 | 188 | 92 |