Assay Details
Gene Symbol: CNMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493004.4 | XP_001493054.1 | 323 | 116 | ||
XM_005601262.3 | XP_005601319.1 | 325 | 116 |

More Information
Set Membership: |
![]() ![]() |
