Assay Details
Gene Symbol: ACOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488399.3 | XP_001488449.1 | 144 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001488399.3 transcripts.
Set Membership: |
![]() ![]() |
