Assay Details
Gene Symbol: LYPD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005601721.3 | XP_005601778.2 | 608 | 62 | ||
XM_014732458.2 | XP_014587944.1 | 768 | 62 | |||
XM_014732459.2 | XP_014587945.1 | 733 | 62 | |||
XM_014732460.2 | XP_014587946.1 | 830 | 62 | |||
XM_014732461.2 | XP_014587947.1 | 839 | 62 | |||
XM_023622856.1 | XP_023478624.1 | 901 | 62 | |||
XM_023622857.1 | XP_023478625.1 | 614 | 62 | |||
XM_023622858.1 | XP_023478626.1 | 1220 | 62 | |||
XM_023622859.1 | XP_023478627.1 | 1149 | 62 | |||
XM_023622860.1 | XP_023478628.1 | 628 | 62 | |||
XM_023622861.1 | XP_023478629.1 | 565 | 62 | |||
XM_023622862.1 | XP_023478630.1 | 462 | 62 | |||
XM_023622863.1 | XP_023478631.1 | 437 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
