Assay Details
Gene Symbol: METTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497963.5 | XP_001498013.1 | 901 | 64 | ||
XM_023623099.1 | XP_023478867.1 | 1002 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
