Assay Details
Gene Symbol: MAP3K20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499196.5 | XP_001499246.5 | 1057 | 66 | ||
XM_005601584.3 | XP_005601641.1 | 1232 | 66 | |||
XM_023622007.1 | XP_023477775.1 | 765 | 66 | |||
XM_023622008.1 | XP_023477776.1 | 1219 | 66 | |||
XM_023622009.1 | XP_023477777.1 | 916 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
