Assay Details
Gene Symbol: CIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014732538.2 | XP_014588024.1 | 345 | 75 | ||
XM_023622023.1 | XP_023477791.1 | 346 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
