Assay Details
Gene Symbol: NFE2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496992.5 | XP_001497042.3 | 527 | 55 | ||
XM_023622057.1 | XP_023477825.1 | 475 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
