Assay Details
Gene Symbol: PRKRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023622066.1 | XP_023477834.1 | 557 | 70 | ||
XM_023622067.1 | XP_023477835.1 | 560 | 70 | |||
XM_023622068.1 | XP_023477836.1 | 665 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
