Assay Details
Gene Symbol: FKBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497400.5 | XP_001497450.3 | 598 | 85 | ||
XM_023622069.1 | XP_023477837.1 | 824 | 85 | |||
GenBank mRNA | JL638548.1 | - | 1 - 2 | 620 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
