Assay Details
Gene Symbol: FLACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014732327.2 | XP_014587813.1 | 1515 | 79 | ||
XM_014732330.2 | XP_014587816.1 | 1284 | 79 | |||
XM_023622356.1 | XP_023478124.1 | 1603 | 79 | |||
XM_023622357.1 | XP_023478125.1 | 1193 | 79 | |||
XM_023622358.1 | XP_023478126.1 | 1604 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
