Assay Details
Gene Symbol: ICOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497868.6 | XP_001497918.3 | 662 | 67 | ||
XM_005601696.3 | XP_005601753.1 | 695 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
