Assay Details
Gene Symbol: PPM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623236.1 | XP_023479004.1 | 294 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023623236.1 transcripts.
Set Membership: |
![]() ![]() |
