Assay Details
Gene Symbol: PPM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623234.1 | XP_023479002.1 | 870 | 93 | ||
XM_023623235.1 | XP_023479003.1 | 865 | 93 | |||
XM_023623236.1 | XP_023479004.1 | 637 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
