Assay Details
Gene Symbol: RPL22L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003363314.3 | XP_003363362.1 | 361 | 112 | ||
XM_005601803.2 | XP_005601860.1 | 331 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
