Assay Details
Gene Symbol: KCNMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494801.5 | XP_001494851.1 | 811 | 60 | ||
XM_005601817.3 | XP_005601874.1 | 919 | 60 | |||
XM_023623330.1 | XP_023479098.1 | 3436 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
