Assay Details
Gene Symbol: DCUN1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014732769.2 | XP_014588255.1 | 839 | 92 | ||
XM_023623370.1 | XP_023479138.1 | 813 | 92 | |||
XM_023623371.1 | XP_023479139.1 | 842 | 92 | |||
GenBank mRNA | JL620640.1 | - | 5 - 6 | 558 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
