Assay Details
Gene Symbol: TRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497821.6 | XP_001497871.2 | 662 | 76 | ||
XM_005601862.3 | XP_005601919.1 | 508 | 76 | |||
XM_014732793.2 | XP_014588279.1 | 374 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
