Assay Details
Gene Symbol: TM4SF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623570.1 | XP_023479338.1 | 476 | 93 | ||
XM_023623571.1 | XP_023479339.1 | 476 | 93 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TM4SF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623570.1 | XP_023479338.1 | 476 | 93 | ||
XM_023623571.1 | XP_023479339.1 | 476 | 93 |