Assay Details
Gene Symbol: TM4SF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623570.1 | XP_023479338.1 | 556 | 90 | ||
XM_023623571.1 | XP_023479339.1 | 556 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
