Assay Details
Gene Symbol: DTX3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623656.1 | XP_023479424.1 | 589 | 85 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
