Assay Details
Gene Symbol: LOC100146890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917281.4 | XP_001917316.1 | 228 | 91 | ||
XM_023623708.1 | XP_023479476.1 | 229 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
