Assay Details
Gene Symbol: POPDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917286.4 | XP_001917321.1 | 2185 | 73 | ||
XM_014732869.2 | XP_014588355.1 | 2190 | 73 | |||
XM_023623709.1 | XP_023479477.1 | 2681 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
