Assay Details
Gene Symbol: NECTIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623803.1 | XP_023479571.1 | 1320 | 79 | ||
XM_023623804.1 | XP_023479572.1 | 1320 | 79 | |||
XM_023623807.1 | XP_023479575.1 | 1028 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
