Assay Details
Gene Symbol: NECTIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623805.1 | XP_023479573.1 | 1610 | 89 | ||
XM_023623806.1 | XP_023479574.1 | 1655 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
