Assay Details
Gene Symbol: HHLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014732902.2 | XP_014588388.1 | 1165 | 170 | ||
XM_023623811.1 | XP_023479579.1 | 1171 | 170 | |||
XM_023623812.1 | XP_023479580.1 | 1506 | 170 | |||
XM_023623813.1 | XP_023479581.1 | 1365 | 170 | |||
XM_023623814.1 | XP_023479582.1 | 1223 | 170 | |||
XM_023623815.1 | XP_023479583.1 | 1248 | 170 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
