Assay Details
Gene Symbol: NXPE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623837.1 | XP_023479605.1 | 1344 | 62 | ||
XM_023623838.1 | XP_023479606.1 | 1515 | 62 | |||
XM_023623839.1 | XP_023479607.1 | 1603 | 62 | |||
XM_023623840.1 | XP_023479608.1 | 1278 | 62 | |||
XM_023623841.1 | XP_023479609.1 | 1347 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
