Assay Details
Gene Symbol: CLDND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023623958.1 | XP_023479726.1 | 414 | 77 | ||
XM_023623959.1 | XP_023479727.1 | 410 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
