Assay Details
Gene Symbol: PLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488028.5 | XP_001488078.1 | 671 | 69 | ||
XM_005607021.3 | XP_005607078.1 | 678 | 69 | |||
XM_023629111.1 | XP_023484879.1 | 675 | 69 |

More Information
Set Membership: |
![]() ![]() |
