Assay Details
Gene Symbol: TTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737438.2 | XP_014592924.2 | 472 | 58 | ||
XM_023629243.1 | XP_023485011.1 | 472 | 58 | |||
XM_023629251.1 | XP_023485019.1 | 486 | 58 | |||
XM_023629260.1 | XP_023485028.1 | 472 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
